Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
Separating funnels, Squibb
Separating funnels, Squibb
Separating funnels, Squibb
Separating funnels, Squibb, made of borosilicate glass 3.3, with NS stopcock plugs and solid lass key. With PE-stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Product Family Options
  • Capacity
    Socket size
  • 50 ml
    19/26 NS
    1 * 2 ST
    1 * 2 ST - 79,30 €
    79,30 €
    1 * 2 ST
       79,30 €
  • 100 ml
    19/26 NS
    1 * 2 ST
    1 * 2 ST - 77,30 €
    77,30 €
    1 * 2 ST
       77,30 €
  • 250 ml
    29/32 NS
    1 * 2 ST
    1 * 2 ST - 87,80 €
    87,80 €
    1 * 2 ST
       87,80 €
  • 500 ml
    29/32 NS
    1 * 2 ST
    1 * 2 ST - 103,00 €
    103,00 €
    1 * 2 ST
       103,00 €
  • 1000 ml
    29/32 NS
    1 * 2 ST
    1 * 2 ST - 136,00 €
    136,00 €
    1 * 2 ST
       136,00 €