Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®

Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®

Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®
Certificates
Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®
Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®

14 Options Available Below

Product Details & Documents

Designed to maximise convenience and efficiency, the ready-to-use Micro Float-A-Lyzer® is ideal for the dialysis of very small sample volumes. Available in two volume sizes, 100 to 200 μl and 400 to 500 μl, the Micro Float-A-Lyzer® features a proprietary biotech grade cellulose ester (CE) membrane incorporated into a pre-assembled, leakproof microdialysis device. Available in 7 concise MWCO’s with colour-coded caps, CE is a synthetic, low-protein binding membrane with no heavy metal and sulfide contaminants. The pre-formed tubular geometry also limits volume increase and sample dilution. The self-standing and self-floating device is designed with a Luer-lok® sample port to provide quick and easy access for loading, in-process testing and total sample recovery using a 1 ml syringe (included). Available in packs of 12, individual units can interlock to form a "flotilla" for the simultaneous dialysis of multiple samples. Only Micro Float-A-Lyzer® and Float-A-Lyzer® G2 assure a 95 to 98% sample recovery, 98% sample purity and <10% sample dilution; all in an easy to use, convenient dialysis device.

  • Colour coded caps
  • Available in two volume sizes
  • Complete with 1 ml syringes

Packaging: Packaged dry with glycerin. Supplied in packs of 12.

Product Family Options

Sample volume sizes 100 to 200 µl

Product Information

  • MWCO
    Colour
    Sample volume
    Availability
    Price
  • 8 - 10 kD
    Yellow
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 0,1 - 0,5 kD
    Green
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 0,5 - 1,0 kD
    Orange
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 3,5 - 5,0 kD
    Black
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 20 kD
    Red
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 50 kD
    White
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 100 kD
    Blue
    0,1 - 0,2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

Sample volume sizes 400 to 500 µl

Product Information

  • MWCO
    Colour
    Sample volume
    Availability
    Price
  • 3,5 - 5,0 kD
    Black
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 50 kD
    White
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 0,1 - 0,5 kD
    Green
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 8 - 10 kD
    Yellow
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 20 kD
    Red
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 100 kD
    Blue
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

  • 0,5 - 1,0 kD
    Orange
    0,4 - 0,5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 194,00 €
    194,00 €
    1 * 12 ST
       194,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.