Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ C18
Solid phase extraction cartridges, HyperSep™ C18
Solid phase extraction cartridges, HyperSep™ C18
Certificates
Solid phase extraction cartridges, HyperSep™ C18
Solid phase extraction cartridges, HyperSep™ C18
12 Options Available Below
Product Details & Documents
These columns feature a highly retentive alkyl-bonded phase for non polar to moderately polar compounds.
  • Hydrophobic reversed phase
  • Retains most organic analytes from aqueous matrices
Product Family Options
Product Information
  • Column Format
    Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • Cartridge
    C18
    40 - 60 µm
    1 ml
    25 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 274,00 €
    274,00 €
    1 * 100 ST
       274,00 €
  • Cartridge
    C18
    40 - 60 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 241,00 €
    241,00 €
    1 * 100 ST
       241,00 €
  • Cartridge
    C18
    40 - 60 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 267,00 €
    267,00 €
    1 * 100 ST
       267,00 €
  • Cartridge
    C18
    40 - 60 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 168,00 €
    168,00 €
    1 * 50 ST
       168,00 €
  • Cartridge
    C18
    40 - 60 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 173,00 €
    173,00 €
    1 * 50 ST
       173,00 €
  • Cartridge
    C18
    40 - 60 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 191,00 €
    191,00 €
    1 * 50 ST
       191,00 €
  • Cartridge
    C18
    40 - 60 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 170,00 €
    170,00 €
    1 * 30 ST
       170,00 €
  • Cartridge
    C18
    40 - 60 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 263,00 €
    263,00 €
    1 * 50 ST
       263,00 €
  • Cartridge
    C18
    40 - 60 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 194,00 €
    194,00 €
    1 * 30 ST
       194,00 €
  • Cartridge
    C18
    40 - 60 µm
    15 ml
    2000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 213,00 €
    213,00 €
    1 * 20 ST
       213,00 €
  • Cartridge
    C18
    40 - 60 µm
    25 ml
    5000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 338,00 €
    338,00 €
    1 * 20 ST
       338,00 €
  • Cartridge
    C18
    40 - 60 µm
    75 ml
    10000 mg
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 274,00 €
    274,00 €
    1 * 10 ST
       274,00 €