ContactUsLinkComponent
Groundwater sampling capsules, Polycap™ GW, Whatman™
undefined
Groundwater sampling capsules, Polycap™ GW, Whatman™
  • Catalog No:
  • 701-0164
  • 701-0173
  • Water flow rate:
  • 60 l/min (1,0 bar)
  • 60 l/min (1,0 bar)
  • Filtration area:
  • 600 cm²
  • 600 cm²
  • Inlet/outlet:
  • 6 to 9 mm stepped barb
  • 6 to 9 mm stepped barb
  • Max. pressure:
  • 4,1 bar
  • 4,1 bar
Frequently Bought Together