Orders LinkContactUsLinkComponent
Drying tubes, U-shaped
Drying tubes, U-shaped
Supplier:   witeg Labortechnik
Drying tubes, U-shaped
Supplier:  witeg Labortechnik
Certificates
Drying tubes, U-shaped
Drying tubes, U-shaped
4 Options Available Below
Product Details & Documents
Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Product Family Options
Product Information
  • Socket size
    Length
    Ø int.
    Availability
    Price
  • 12/21 NS
    100 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,80 €
    55,80 €
    1 * 1 ST
       55,80 €
  • 14/23 NS
    125 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,40 €
    61,40 €
    1 * 1 ST
       61,40 €
  • 19/26 NS
    150 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • 19/26 NS
    250 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €