Orders LinkContactUsLinkComponent
Automatic burettes, Schilling pattern
Automatic burettes, Schilling pattern
Supplier:   Poulten & Graf
Automatic burettes, Schilling pattern
Supplier:  Poulten & Graf
Certificates
Automatic burettes, Schilling pattern
Automatic burettes, Schilling pattern
9 Options Available Below
Product Details & Documents
AR-GLAS®, clear or amber, class B.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, reduced distance between subdivision marks
  • Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
Error limits according to class B, DIN EN ISO 385
Certifications: Error limits according to class B, DIN EN ISO 385
Product Family Options
Clear glass
Product Information
  • Capacity
    Division
    Availability
    Price
  • 5 ml
    0,05 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 10 ml
    0,05 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 15 ml
    0,10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 25 ml
    0,10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 120,00 €
    120,00 €
    1 * 1 ST
       120,00 €
  • 50 ml
    0,10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
Amber glass
Product Information
  • Capacity
    Division
    Availability
    Price
  • 10 ml
    0,05 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • 15 ml
    0,10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • 25 ml
    0,10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €
  • 50 ml
    0,10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €