Orders LinkContactUsLinkComponent
Swabs
Swabs
Swabs
Supplier:  LP ITALIANA
Certificates
Swabs
8 Options Available Below
Product Details & Documents
Sterile and non sterile swabs designed for use in microbiology.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Shaft/head
    Ø
    Length
    Availability
    Price
  • LPIT112191
    Non sterile swabs
    Non sterile swabs
    Wood/cotton
    2,2 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 104,00 €
    104,00 €
    1 * 3.000 ST
       104,00 €
  • LPIT112491
    Non sterile swabs
    Non sterile swabs
    Plastic/cotton
    2,2 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 221,00 €
    221,00 €
    1 * 3.000 ST
       221,00 €
  • LPIT112591
    Non sterile swabs
    Non sterile swabs
    Plastic/rayon
    2,2 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 247,00 €
    247,00 €
    1 * 3.000 ST
       247,00 €
  • LPIT112598
    Sterile swabs (single wrapped)
    Sterile swabs (single wrapped)
    Plastic/rayon
    2,2 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 204,00 €
    204,00 €
    1 * 1.000 ST
       204,00 €
  • LPIT112691
    Non sterile swabs
    Non sterile swabs
    Aluminium/cotton
    1 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 1 150,00 €
    1 150,00 €
    1 * 3.000 ST
       1 150,00 €
  • LPIT112698
    Sterile swabs (single wrapped)
    Sterile swabs (single wrapped)
    Aluminium/cotton
    1 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 528,00 €
    528,00 €
    1 * 1.000 ST
       528,00 €
  • LPIT112298
    Sterile swabs (single wrapped)
    Sterile swabs (single wrapped)
    Wood/cotton
    2,2 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 164,00 €
    164,00 €
    1 * 1.000 ST
       164,00 €
  • LPIT112498
    Sterile swabs (single wrapped)
    Sterile swabs (single wrapped)
    Plastic/cotton
    2,2 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 195,00 €
    195,00 €
    1 * 1.000 ST
       195,00 €