Orders LinkContactUsLinkComponent
Drying tubes, bend
Drying tubes, bend
Supplier:   witeg Labortechnik
Drying tubes, bend
Supplier:  witeg Labortechnik
Certificates
Drying tubes, bend
Drying tubes, bend
2 Options Available Below
Product Details & Documents
Borosilicate glass 3.3. Angled at 75° with ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Product Family Options
Product Information
  • Cone size
    Availability
    Price
  • 24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,80 €
    26,80 €
    1 * 1 ST
       26,80 €
  • 19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €