ContactUsLinkComponent
Volumetric flasks, class B, white markings
Volumetric flasks, class B, white markings
Catalog # HECH42089092
undefined
Volumetric flasks, class B, white markings
Catalog # HECH42089092
Specifications
  • NS:
    -
  • Capacity:
    3000 ml
  • Seq:
    01
  • Pk:
    1
  • Tolerance:
    1,2 ± ml
Specifications
About this item
Borosilicate glass 3.3.
  • Higher accuracy than standard class B, Tolerance: 1,5 times of the official tolerance of class A
  • Calibrated to contain (marked 'In')
*Wide neck
Certifications: In accordance with DIN EN ISO 1042