Orders LinkContactUsLinkComponent
Containers with pressure cap
Containers with pressure cap
Supplier:   LP ITALIANA
Containers with pressure cap
Supplier:  LP ITALIANA
Certificates
Containers with pressure cap
Containers with pressure cap
2 Options Available Below
Product Details & Documents
Container made of PS, clear, with PE snap cap.
  • Good chemical resistance
  • Lightweight
  • Easy to clean
Ideal for collecting, storing and transporting samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Cap colour
    Version
    Material
    Availability
    Price
  • 30 ml
    34 mm
    41 mm
    Yellow
    sterile (ionising radiation)
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 359,00 €
    359,00 €
    1 * 1.000 ST
       359,00 €
  • 45 ml
    34 mm
    60 mm
    White
    non sterile
    PS
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 166,00 €
    166,00 €
    1 * 750 ST
       166,00 €