ContactUsLinkComponent
Volumetric flasks, class A, trapezoidal shape, blue markings
Volumetric flasks, class A, trapezoidal shape, blue markings
Catalog # HECH42110024
undefined
Volumetric flasks, class A, trapezoidal shape, blue markings
Catalog # HECH42110024
Specifications
  • NS:
    7/16
  • Capacity:
    10 ml
  • Pk:
    1
  • Tolerance:
    0,025 ± ml
Specifications
About this item
DURAN® Borosilicate glass 3.3 with PP stopper.
  • Calibrated to contain (TC, In)
  • Blue markings
  • Conformity certified (DE-M marking)
The base is considerably larger than of standard volumetric flasks with same volume, thus improved stability.
In accordance with DIN EN ISO 1042