Orders LinkContactUsLinkComponent
Original Perfusor® syringes
Original Perfusor® syringes
Supplier:  B. Braun
Certificates
Original Perfusor® syringes
Original Perfusor® syringes
4 Options Available Below
Product Details & Documents
PP syringe with safe plunger backstop and minimal residual volume.
  • Oval grip plate for handling of the syringe and for prevention of rolling
  • Plunger stopper with double sealing ring for slow aspiration or infusion of drug solutions, made of synthetic material or natural latex-free
  • Centric Luer Lock cone for the attachment of a needle or infusion tubing
Product Family Options
Product Information
  • Capacity
    Graduation
    Length
    Ext.Ø
    Type
    Availability
    Price
  • BRAU8728810F-06
    3-Piece syringe for infusion pumps, with aspiration needle, transparent
    3-Piece syringe for infusion pumps, with aspiration needle, transparent
    50 ml
    1,00 ml
    30 mm
    2 mm
    Centric cone
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 178,00 €
    178,00 €
    1 * 100 ST
       178,00 €
  • 613-0120
    3-Piece syringe for infusion pumps, without aspiration needle, transparent
    3-Piece syringe for infusion pumps, without aspiration needle, transparent
    50 ml
    — ml
    — ml
    — mm
    Centric cone
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 178,00 €
    178,00 €
    1 * 100 ST
       178,00 €
  • BRAU8728852F-06
    Original Perfusor® Syringe 50 ml, with particle filter cannula
    Original Perfusor® Syringe 50 ml, with particle filter cannula
    50 ml
    — ml
    — ml
    — mm
    Centric cone
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 213,00 €
    213,00 €
    1 * 100 ST
       213,00 €
  • BRAU8728861F-06
    Original Perfusor® syringe 50 ml, with particle filter cannula, type UV-Protect, orange-transparent
    Original Perfusor® syringe 50 ml, with particle filter cannula, type UV-Protect, orange-transparent
    50 ml
    — ml
    — ml
    — mm
    Centric cone
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 498,00 €
    498,00 €
    1 * 100 ST
       498,00 €