Orders LinkContactUsLinkComponent
Nuts, hexagonal, BOLA
Nuts, hexagonal, BOLA
Supplier:   Bohlender
Nuts, hexagonal, BOLA
Supplier:  Bohlender
Certificates
Nuts, hexagonal, BOLA
Nuts, hexagonal, BOLA
4 Options Available Below
Product Details & Documents
PTFE, white.
  • Temperature resistance: from -200 °C to +250 °C
  • Universal chemical resistance
  • Similar to DIN 934 / DIN EN ISO 4032
Product Family Options
Product Information
  • Thread
    Height
    Head Ø
    Pitch
    Material
    Availability
    Price
  • M 8
    9,0 mm
    13 mm
    1,25 mm
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,70 €
    20,70 €
    1 * 1 ST
       20,70 €
  • M 6
    7,5 mm
    10 mm
    1,0 mm
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,70 €
    12,70 €
    1 * 1 ST
       12,70 €
  • M 5
    6,0 mm
    8 mm
    0,8 mm
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,70 €
    11,70 €
    1 * 1 ST
       11,70 €
  • M 4
    5,0 mm
    7 mm
    0,7 mm
    PTFE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €