Screws with countersunk, BOLA
Supplier: Bohlender
Aller à :
À propos de cet article
PTFE, white.
- Temperature resistance: from -200 °C to +250 °C
- Universal chemical resistance
- FDA conform
- Similar to DIN 963/DIN EN ISO 2009
4 Options disponibles ci-dessous
Product Details & Documents
PTFE, white.
- Temperature resistance: from -200 °C to +250 °C
- Universal chemical resistance
- FDA conform
- Similar to DIN 963/DIN EN ISO 2009
Product Family Options
Informations sur le produit
- Head ØThreadPitchMaterialUsable lengthAvailabilityPrice