ContactUsLinkComponent
Flasks, pear shaped, Claisen-Vigreux, with two necks
Flasks, pear shaped, Claisen-Vigreux, with two necks
Catalog # WITG0.786.025
Supplier:  witeg Labortechnik
undefined
Flasks, pear shaped, Claisen-Vigreux, with two necks
Catalog # WITG0.786.025
Supplier:  witeg Labortechnik
Specifications
  • Capacity:
    25 ml
  • Pk:
    1
  • Side necks:
    14/23 NS
  • Centre neck:
    14/23 NS
Specifications
About this item
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Incorporates a short, Vigreux type, fractionating column. Column indents aid intimate contact between vapour and liquid.