ContactUsLinkComponent
Flasks, pear shaped, acc. to Claisen, with two necks
Flasks, pear shaped, acc. to Claisen, with two necks
Catalog # WITG0.783.025
Supplier:  witeg Labortechnik
undefined
Flasks, pear shaped, acc. to Claisen, with two necks
Catalog # WITG0.783.025
Supplier:  witeg Labortechnik
Specifications
  • Capacity:
    25 ml
  • Pk:
    1
  • Side necks:
    14/23 NS
  • Centre neck:
    14/23 NS
Specifications
About this item
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Center necks with socked NS 14/23, side neck with cone NS 14/23.