ContactUsLinkComponent
Leak detection additives, Reveal™
Leak detection additives, Reveal™
Catalog # UVPA96-0150-03
undefined
Leak detection additives, Reveal™
Catalog # UVPA96-0150-03
Specifications
  • Description:
    A-680 Plus additive for oil, fuel and ATF systems
  • Seq:
    08
  • Pk:
    1
Specifications
About this item
This leak detection system brings leaks to light with an ultraviolet lamp and specially formulated fluorescent tracing additives.
  • Locating all leaks at once, even multiple small leaks in the same system
  • Tracer additives work easily and quickly in any system using water, oil, ATF, fuel or refrigerant