Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acriflavine hydrochloride
Acriflavine hydrochloride
Acriflavine hydrochloride
ROTH9621.1
:
Roth Carl
:
Acriflavine hydrochloride
ROTH9621.1
:
Roth Carl
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
578.36 g/mol
:
8063-24-9
:
C₂₇H₂₈Cl₄N₆
:
MFCD00069039