Order Entry
Belgium
ContactUsLinkComponent
Horseradish peroxidase
Horseradish peroxidase
Supplier:  Serva
Certificates
Horseradish peroxidase
Supplier:  Serva

Specifications

  • CAS No.:
    9003-99-0
  • Species:
    Horseradish
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00071339
  • Enzyme name:
    Peroxidase

Specifications

Frequently Bought Together