Order Entry
Belgium
Orders LinkContactUsLinkComponent
E-64

E-64

Marketsource
Supplier:   Serva
E-64
Supplier:  Serva
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SERA21100.01
  • SERA21100.02
  • Protein/peptide name:
  • E-64
  • E-64
  • Protein synonyms:
  • 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
  • 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
  • CAS No.:
  • 66701-25-5
  • 66701-25-5
  • MDL No.:
  • MFCD00080261
  • MFCD00080261
  • Molecular formula:
  • C₁₅H₂₇N₅O₅
  • C₁₅H₂₇N₅O₅
  • Storage conditions:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 102,00 €
    102,00 €
    1 * 5 mg
       102,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    SERA21100.01
    Protein/peptide name
    E-64
    Protein synonyms
    3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
    CAS No.
    66701-25-5
    MDL No.
    MFCD00080261
    Molecular formula
    C₁₅H₂₇N₅O₅
    Storage conditions
    Freezer
    Marketsource
  • Glass bottle for solids
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 363,00 €
    363,00 €
    1 * 25 mg
       363,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    SERA21100.02
    Protein/peptide name
    E-64
    Protein synonyms
    3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
    CAS No.
    66701-25-5
    MDL No.
    MFCD00080261
    Molecular formula
    C₁₅H₂₇N₅O₅
    Storage conditions
    Freezer
    Marketsource