E-64
5 mg
Catalog # 786-049
Supplier: G-Biosciences
Jump to:
Product Details & Documents
E-64 is an irreversible inhibitor of cysteine proteases. It interacts with the Sn subsites of proteases but has no action on cysteine residues in other proteins.
Specifications
Specifications
- Pk:5 mg
- Cat. No.:786-049
- Molecular formula:C₁₅H₂₇N₅O₅
- Protein/peptide name:E-64
- Storage conditions:Freezer
- Protein synonyms:3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
- MDL No.:MFCD00080261
- CAS No.:66701-25-5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



