Order Entry
Belgium
ContactUsLinkComponent
VWR®, Malt extract, bacteriological grade
VWR®, Malt extract, bacteriological grade
Catalog # J873-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Malt extract, bacteriological grade
Catalog # J873-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Description:
    Malt extract, bacteriological grade
  • Seq:
    02
  • Solubility:
    6% in water
  • Storage temperature:
    Cold
  • pH:
    6% water, 4,5 to 5,7

Specifications

About this item

Used for the preparation of media for the detection of yeasts and moulds.