ContactUsLinkComponent
VWR®, dTTP solution
VWR®, dTTP solution
Catalog # E542-40UM
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, dTTP solution
Catalog # E542-40UM
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    40 µmol
  • Description:
    dTTP solution, 100 mM solution, pH 7,0
Specifications
About this item
Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA.
  • Building blocks for the amplification of DNA
  • High purity grade
DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule.