Order Entry
Belgium
ContactUsLinkComponent
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%, Ultra Pure Grade
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%, Ultra Pure Grade
Supplier:  VWR Chemicals
Certificates
undefined
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%, Ultra Pure Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • A250/A260:
    0.75 – 0.81
  • A280/A260:
    0.14 – 0.18
  • AMP & ADP:
    ≤0.5%
  • Em (259 nm,Phosphate Buffer,pH 7.0):
    ≥14700
  • Heavy Metals:
    ≤0.003%
  • Moisture (KF):
    <10%
  • Purity:
    >98%
  • Sodium (Flame Photom):
    7 – 8%
  • Solubility (5%, Water):
    pass

Specification Test Results

Related Information

Frequently Bought Together