Order Entry
Belgium
ContactUsLinkComponent
2'-Deoxyadenosine monohydrate
2'-Deoxyadenosine monohydrate
Catalog # MOLE34645670-5G
Supplier:  Molekula
CAS Number:  
2'-Deoxyadenosine monohydrate
Catalog # MOLE34645670-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :269.26 g/mol (251.24 anhy)
Storage Temperature:Ambient
EINECS:213-488-7
CAS Number:16373-93-6
Formula:C₁₀H₁₃N₅O₃·1H₂O
MDL Number:MFCD00149364