
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)
Catalog # SERA12310.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:616.49 g/mol
EINECS:213-903-1
:GHS07
Storage Temperature:Ambient
CAS Number:1064-48-8
Formula:C₂₂H₁₄N₆Na₂O₉S₂
MDL Number:MFCD00004017