Erythromycin
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: (-)-Erythromycin
- CAS Number:114-07-8
- Formula:C₃₇H₆₇NO₁₃
- MDL Number:MFCD00084654
- Molecular Weight:733.95 g/mol
- Storage Temperature:Ambient
- EINECS:204-040-1
2 Options Available Below
Product Details & Documents
Synonyms: (-)-Erythromycin
- CAS Number:114-07-8
- Formula:C₃₇H₆₇NO₁₃
- MDL Number:MFCD00084654
- Molecular Weight:733.95 g/mol
- Storage Temperature:Ambient
- EINECS:204-040-1
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



