Order Entry
Belgium
Orders LinkContactUsLinkComponent
Erythromycin

Erythromycin

Glass bottle for solids, 5 g
Catalog # MOLE28466420-5G
Irritant
Marketsource
Supplier:   Molekula
Erythromycin
Catalog # MOLE28466420-5G
Supplier:  Molekula
Marketsource
CAS Number:  114-07-8
Erythromycin

Glass bottle for solids, 5 g

CAS Number:  114-07-8
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (-)-Erythromycin
  • CAS Number:114-07-8
  • Formula:C₃₇H₆₇NO₁₃
  • MDL Number:MFCD00084654
  • Molecular Weight:733.95 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-040-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications