
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # ROTH3745.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:389.381 g/mol
EINECS:222-042-0
:GHS07
Storage Temperature:Refrigerator
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063