Order Entry
Belgium
ContactUsLinkComponent
Chicken Native Avidin (from Egg)
Chicken Native Avidin (from Egg)
Certificates
Chicken Native Avidin (from Egg)

Specifications

  • CAS No.:
    1405-69-2
  • Protein/peptide type:
    Native
  • Source:
    Egg
  • Species:
    Chicken
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00130572
  • Protein/peptide name:
    Avidin

Specifications

Frequently Bought Together