Synperonic® F68
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), PEG-PPG-PEG, Kolliphor® P 188, Synperonic® F68, Lutrol® F68, Pluronic® F-68
- CAS Number:9003-11-6
- Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
- MDL Number:MFCD00082049
- Molecular Weight:8400
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), PEG-PPG-PEG, Kolliphor® P 188, Synperonic® F68, Lutrol® F68, Pluronic® F-68
- CAS Number:9003-11-6
- Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
- MDL Number:MFCD00082049
- Molecular Weight:8400
- Storage Temperature:Ambient
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



