Order Entry
Belgium
Orders LinkContactUsLinkComponent
Chymostatin

Chymostatin

Marketsource
Supplier:   Serva
Chymostatin
Supplier:  Serva
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SERA17158.01
  • SERA17158.02
  • Protein/peptide name:
  • Chymostatin
  • Chymostatin
  • CAS No.:
  • 9076-44-2
  • 9076-44-2
  • MDL No.:
  • MFCD00071059
  • MFCD00071059
  • Storage conditions:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    SERA17158.01
    Protein/peptide name
    Chymostatin
    CAS No.
    9076-44-2
    MDL No.
    MFCD00071059
    Storage conditions
    Freezer
    Marketsource
  • Glass bottle for solids
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    SERA17158.02
    Protein/peptide name
    Chymostatin
    CAS No.
    9076-44-2
    MDL No.
    MFCD00071059
    Storage conditions
    Freezer
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.