Bacitracin
Marketsource
Supplier: Serva
Jump to:
About this item
- CAS Number:1405-87-4
- Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL Number:MFCD00062640
- Molecular Weight:1421.60 g/mol
2 Options Available Below
Product Details & Documents
- CAS Number:1405-87-4
- Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL Number:MFCD00062640
- Molecular Weight:1421.60 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



