Order Entry
Belgium
ContactUsLinkComponent
Bovine aprotinin (from Lung)
Bovine aprotinin (from Lung)
Supplier:  Serva
Certificates
Bovine aprotinin (from Lung)
Supplier:  Serva

Specifications

  • CAS No.:
    9087-70-1
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00130541
  • Protein synonyms:
    Trypsin inhibitor, pancreatic basic
  • Protein/peptide name:
    Aprotinin
  • Molecular formula:
    C₂₈₄H₄₃₂N₈₄O₇₉S₇
  • Source:
    Lung

Specifications

Frequently Bought Together