Order Entry
Belgium
ContactUsLinkComponent
Aprotinin (Trypsin inhibitor, pancreatic basic) (from bovine lung), proteomics grade
Aprotinin (Trypsin inhibitor, pancreatic basic) (from bovine lung), proteomics grade
Catalog # M167-10MG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Aprotinin (Trypsin inhibitor, pancreatic basic) (from bovine lung), proteomics grade
Catalog # M167-10MG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    9087-70-1
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00130541
  • Protein synonyms:
    Trypsin inhibitor, pancreatic basic
  • Protein/peptide name:
    Aprotinin
  • Grade:
    Proteomics Grade
  • Molecular formula:
    C₂₈₄H₄₃₂N₈₄O₇₉S₇

Specifications

About this item

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.

Activity (KIU/mg)      ≤4500

Purity (HPLC)      Pass