
Nonidet® P 40 Substitute (NP-40), Millipore®
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Qualitative Filter Papers
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
2,2,2-Trichloroethanol ≥99%
: Thermo Fisher Scientific
Cartridges for deionisers
: Thermo Fisher Scientific
SURE competent cells
: AGILENT
IGEPAL® CA-630
: Thermo Fisher Scientific