Order Entry
Belgium
ContactUsLinkComponent
Trihexyl(tetradecyl)phosphonium chloride ≥93% (CYPHOS® IL 101)
Irritant
Trihexyl(tetradecyl)phosphonium chloride ≥93% (CYPHOS® IL 101)
Catalog # STRE15-6382
CAS Number:  
Irritant
Trihexyl(tetradecyl)phosphonium chloride ≥93% (CYPHOS® IL 101)
Catalog # STRE15-6382
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :519.31 g/mol
Density:0,894
 :GHS07
CAS Number:258864-54-9
Formula:C₃₂H₆₈ClP
MDL Number:MFCD04112746