
Cetyldimethylethylammonium bromide
Catalog # MOLE30028350-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:378.4741 g/mol
Storage Temperature:Ambient
EINECS:204-672-8
UN:2811
ADR:6.1,III
:GHS07
CAS Number:124-03-8
Formula:C₂₀H₄₄BrN
MDL Number:MFCD00011774