Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Linamarase (beta-D-Glucosidase)
Linamarase (beta-D-Glucosidase)
Linamarase (beta-D-Glucosidase)
Catalog # MEGAE-BGLUC
Supplier:
MEGAZYME
CAS Number:
Expand image
Linamarase (beta-D-Glucosidase)
Catalog # MEGAE-BGLUC
Supplier:
MEGAZYME
CAS Number:
Specifications
Pk:
200
CAS No.:
9001-22-3
Enzyme name:
Linamarase (beta-D-Glucosidase)
Pk Info:
200 U
Specifications
About this item
Product Details
Product Details