Ammonium chloride 2 mol/l (2 N) in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:12125-02-9
:
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Storage Temperature:Ambient
EINECS:235-186-4
Frequently Bought Together