Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pepsin
Pepsin
Pepsin
DAKOS300230
:
Dako
:
Pepsin
DAKOS300230
:
Dako
:
Pk:
6
CAS No.:
9001-75-6
Storage conditions:
Refrigerator
Enzyme name:
Pepsin
Pk Info:
6×2 g