Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB), pure

3,3',5,5'-Tetramethylbenzidine (TMB), pure

3,3',5,5'-Tetramethylbenzidine (TMB), pure
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB), pure

2 Options Available Below

Product Details & Documents

Use as water soluble, non-carcinogenic peroxidase substrate for ELISA application. Further applications: detection of cholesterol in blood serum, and blood in urine. Measure absorbance of the yellow reaction product at 450 nm.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:166…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g