Order Entry
Belgium
ContactUsLinkComponent
Thallium standard solution, 1000 mg/L Tl in dil. nitric acid (from TlNO3) standard for AAS and ICP
Thallium standard solution, 1000 mg/L Tl in dil. nitric acid (from TlNO3) standard for AAS and ICP
Catalog # PERCN9300170
Supplier:  PerkinElmer
CAS Number:  
Thallium standard solution, 1000 mg/L Tl in dil. nitric acid (from TlNO3) standard for AAS and ICP
Catalog # PERCN9300170
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Thallium
  • Symbol:
    Tl

Specifications

About this item