Order Entry
Belgium
ContactUsLinkComponent
Silicon standard solution, 1,000 mg/L Si in water (from (NH4)2SiF6) standard for AAS and ICP
Silicon standard solution, 1,000 mg/L Si in water (from (NH4)2SiF6) standard for AAS and ICP
Catalog # PERCN9300150
Supplier:  PerkinElmer
CAS Number:  
Silicon standard solution, 1,000 mg/L Si in water (from (NH4)2SiF6) standard for AAS and ICP
Catalog # PERCN9300150
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item