Order Entry
Belgium
ContactUsLinkComponent
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid (from Lu₂O₃) standard for AAS and ICP
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid (from Lu₂O₃) standard for AAS and ICP
Catalog # PERCN9300130
Supplier:  PerkinElmer
CAS Number:  
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid (from Lu₂O₃) standard for AAS and ICP
Catalog # PERCN9300130
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Lutetium
  • Symbol:
    Lu

Specifications

About this item