Order Entry
Belgium
ContactUsLinkComponent
Erbium Standard Solution, 1000 mg/L Er in dil. Nitric Acid (from Er₂O₃) Standard for AAS and ICP
Erbium Standard Solution, 1000 mg/L Er in dil. Nitric Acid (from Er₂O₃) Standard for AAS and ICP
Catalog # PERCN9300116
Supplier:  PerkinElmer
CAS Number:  
Erbium Standard Solution, 1000 mg/L Er in dil. Nitric Acid (from Er₂O₃) Standard for AAS and ICP
Catalog # PERCN9300116
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Erbium
  • Symbol:
    Er

Specifications

About this item