Order Entry
Belgium
ContactUsLinkComponent
 
Iron standard solution, 1000 mg/L Fe in dil. nitric acid (from Fe) standard for AAS and ICP
Catalog # PERCN9300126
Supplier:  PerkinElmer
CAS Number:  
Iron standard solution, 1000 mg/L Fe in dil. nitric acid (from Fe) standard for AAS and ICP
Catalog # PERCN9300126
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item