Order Entry
Belgium
ContactUsLinkComponent
Yttrium standard solution, 1000 mg/L Y in dil. nitric acid (from Y₂O₃) standard for AAS and ICP
Yttrium standard solution, 1000 mg/L Y in dil. nitric acid (from Y₂O₃) standard for AAS and ICP
Catalog # PERCN9300167
Supplier:  PerkinElmer
CAS Number:  
Yttrium standard solution, 1000 mg/L Y in dil. nitric acid (from Y₂O₃) standard for AAS and ICP
Catalog # PERCN9300167
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item