Order Entry
Belgium
ContactUsLinkComponent
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) standard for AAS and ICP
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) standard for AAS and ICP
Catalog # PERCN9300163
Supplier:  PerkinElmer
CAS Number:  
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) standard for AAS and ICP
Catalog # PERCN9300163
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1
  • Element:
    Tungsten
  • Symbol:
    W

Specifications

About this item