Strontium standard solution, 1000 mg/L Sr in dil. nitric acid (from Sr(NO₃)₂) standard for AAS and ICP
Catalog # PERCN9300153
Fournisseur: PerkinElmer
Numéro CAS:
Spécifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pk:1
- Element:Strontium
- Symbol:Sr